3-Acetamido-5-carboxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

A584665-1G A584665-5G
77884-418EA 212 USD
77884-418 77884-419
3-Acetamido-5-carboxyphenylboronic acid ≥98%
3-Acetamido-5-carboxyphenylboronic acid

Caution: for Research Use Only

Formula: C₉H₁₀BNO₅
MDL Number: MFCD08436066
CAS Number: 108749-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR