Xylenol orange tetrasodium salt Indicator

Supplier: Ambeed
Warning

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

A579321-25G A579321-100G
77617-436EA 110.21 USD
77617-436 77749-192
Xylenol orange tetrasodium salt Indicator
Xylenol orange tetrasodium salt

Caution: for Research Use Only

Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210
MDL Number: MFCD00044293
CAS Number: 3618-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR