Tri-tert-butyl 2,2',2''-(10-(2-((2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate ≥95%

Supplier: Ambeed
Warning

A562779-100MG A562779-50MG A562779-250MG A562779-1G
77747-157EA 41 USD
77747-157 77747-160 77747-159 77747-158
Tri-tert-butyl 2,2',2''-(10-(2-((2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate ≥95%
Tri-tert-butyl 2,2',2''-(10-(2-((2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

Caution: for Research Use Only

Formula: C₃₂H₅₅N₅O₁₀
MDL Number: MFCD26131244
CAS Number: 819869-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR