9-Hydroxy-10-methoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride ≥98%
Supplier: Ambeed
|
Warning
|
A531013-100MG
A531013-1G
A531013-250MG
A531013-50MG
77613-662EA
228.66
USD
77613-662
77901-688
77900-893
77900-659
9-Hydroxy-10-methoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride ≥98%
9-Hydroxy-10-methoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride
Caution: for Research Use Only
Formula:
C₁₉H₁₆ClNO₄ |
MDL Number:
MFCD01658905 CAS Number: 15401-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...