4-Bromo-2-ethylbenzoic acid ≥95%

Supplier: Ambeed
Warning

A528798-1G A528798-25G
77686-362EA 45 USD
77686-362 77901-294
4-Bromo-2-ethylbenzoic acid ≥95%
4-Bromo-2-ethylbenzoic acid

Caution: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD18410376
CAS Number: 644984-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR