(4-Nitrophenyl)-α-D-glucopyranoside 98
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside
A509900-5G
A509900-25G
A509900-250MG
77617-180EA
92.7
USD
77617-180
77749-043
77749-042
(4-Nitrophenyl)-α-D-glucopyranoside 98
(4-Nitrophenyl) α-D-glucopyranoside
Caution: for Research Use Only
Formula:
C₁₂H₁₅NO₈ MW: 301.25 g/mol |
MDL Number:
MFCD00064088 CAS Number: 3767-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...