EDTA tetrasodium salt hydrate ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethylenediaminetetraacetic acid tetrasodium salt hydrate
, Edetic acid tetrasodium salt hydrate
, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate
, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate
, Tetrasodium ethylenediaminetetraacetate hydrate
, Kestranal® 4S hydrate
A509330-1000G
A509330-500G
A509330-100G
77594-596EA
15.38
USD
77594-596
77602-176
77602-174
EDTA tetrasodium salt hydrate ≥98%
EDTA tetrasodium salt hydrate
Caution: for Research Use Only
Formula:
C₁₀H₁₂N₂O₈Na₄·xH₂O MW: 380.17 g/mol (anhy) |
MDL Number:
MFCD00150025 CAS Number: 13235-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...