1-(3-Dimethylaminopropyl)piperazine ≥95%

Supplier: Ambeed
Warning

A498475-1G A498475-5G A498475-10G A498475-250MG A498475-25G
77626-990EA 31.93 USD
77626-990 77626-992 77878-271 77878-272 77878-273
1-(3-Dimethylaminopropyl)piperazine ≥95%
1-(3-Dimethylaminopropyl)piperazine

Caution: for Research Use Only

Formula: C₉H₂₁N₃
MDL Number: MFCD00082601
CAS Number: 877-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR