(S)-3,3'-Bis(10-phenyl-9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate ≥98%

Supplier: Ambeed
Warning

A491662-50MG A491662-250MG
77594-540EA 177.33 USD
77594-540 77747-009
(S)-3,3'-Bis(10-phenyl-9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate ≥98%
(S)-3,3'-Bis(10-phenyl-9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate

Caution: for Research Use Only

Formula: C₆₀H₃₇O₄P
CAS Number: 2207600-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR