6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥97%
Supplier: Ambeed
|
Warning
|
A470567-250MG
A470567-100MG
77572-480EA
490
USD
77572-480
77602-048
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥97%
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)
Caution: for Research Use Only
Formula:
C₂₅H₁₅NO₉ |
MDL Number:
MFCD00077323 CAS Number: 92557-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...