Cyclohexanaminium 4-(1-(4-hydroxyphenyl)-3-oxo-1,3-dihydroisobenzofuran-1-yl)phenyl phosphate ≥85%

Supplier: Ambeed
Warning

A473317-100MG A473317-1G A473317-250MG A473317-5G
77602-056EA 11.28 USD
77602-056 77602-058 77602-060 77669-122
Cyclohexanaminium 4-(1-(4-hydroxyphenyl)-3-oxo-1,3-dihydroisobenzofuran-1-yl)phenyl phosphate ≥85%
Cyclohexanaminium 4-(1-(4-hydroxyphenyl)-3-oxo-1,3-dihydroisobenzofuran-1-yl)phenyl phosphate

Caution: for Research Use Only

Formula: C₃₂H₄₁N₂O₇P
MDL Number: MFCD00070132
CAS Number: 14815-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR