Furan-2-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Furan-2-sulfonylchloride, Furan-2-sulphonylchloride

A469775-100MG A469775-1G A469775-250MG
77880-021EA 75 USD
77880-021 77880-022 77880-023
Furan-2-sulfonyl chloride ≥98%
Furan-2-sulfonyl chloride

Caution: for Research Use Only

Formula: C₄H₃ClO₃S
MDL Number: MFCD06658968
CAS Number: 52665-48-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR