EDTA dipotassium salt dihydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrateEthylenedinitrilotetra-acetic acid dipotassium salt dihydrateEthylenedinitrilotetraacetic acid dipotassium saltTitriplex® Potassium saltEdetic acid dipotassium salt dihydrateEDTA dipotassium dihydrateDipotassium dihydrogen ethylenediaminetetraacetate dihydratePotassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrateEthylenediaminetetra-acetic acid dipotassium salt dihydrate

A433353-500G
77601-874EA 47.15 USD
77601-874
EDTA dipotassium salt dihydrate ≥98%
EDTA dipotassium salt dihydrate

Caution: for Research Use Only

Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
MDL Number: MFCD00150036
CAS Number: 25102-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR