2-Amino-5-fluoronicotinic acid ≥97%
Supplier: Ambeed
|
Warning
|
A416451-1G
A416451-250MG
77884-080EA
200
USD
77884-080
77884-081
2-Amino-5-fluoronicotinic acid ≥97%
2-Amino-5-fluoronicotinic acid
Caution: for Research Use Only
Formula:
C₆H₅FN₂O₂ MW: 156.12 g/mol |
MDL Number:
MFCD13190373 CAS Number: 1196154-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...