2,5-Dioxopyrrolidin-1-yl hex-5-ynoate ≥98%
Supplier: Ambeed
|
Warning
|
A400731-1G
A400731-250MG
A400731-5G
77601-762EA
181.43
USD
77601-762
77601-764
77601-766
2,5-Dioxopyrrolidin-1-yl hex-5-ynoate ≥98%
2,5-Dioxopyrrolidin-1-yl hex-5-ynoate
Caution: for Research Use Only
Formula:
C₁₀H₁₁NO₄ |
MDL Number:
MFCD28898858 CAS Number: 906564-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...