MK-571 sodium salt ≥95%

Supplier: Ambeed
Warning

Synonyms: Sodium (E)-3-(((3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)((3-(dimethylamino)-3-oxopropyl)thio)methyl)thio)propanoate, MK-571 sodium

A407864-100MG A407864-50MG
77886-091EA 471 USD
77886-091 77886-092
MK-571 sodium salt ≥95%
MK-571 sodium salt

Caution: for Research Use Only

Formula: C₂₆H₂₆ClN₂O₃S₂·Na
MDL Number: MFCD11114392
CAS Number: 115103-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR