2,2,2-Trifluoroethyl acrylate ≥95% stabilized

Supplier: Ambeed
Danger

Synonyms: 2,2,2-TrifluoroethylacrylateAcrylic acid 2,2,2-trifluoroethyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A387394-25G
77572-164EA 39 USD
77572-164
2,2,2-Trifluoroethyl acrylate ≥95% stabilized
2,2,2-Trifluoroethyl acrylate

Caution: for Research Use Only

Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
MDL Number: MFCD00000444
CAS Number: 407-47-6
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR