Decacarbonyldirhenium ≥98%

Supplier: Ambeed
Danger

Synonyms: Rhenium carbonyl

A392186-250MG A392186-100MG A392186-1G
77594-198EA 62.53 USD
77594-198 77613-184 77613-186
Decacarbonyldirhenium ≥98%
Decacarbonyldirhenium

Caution: for Research Use Only

Formula: C₁₀O₁₀Re₂
MW: 652.52
Density: 2.87
MDL Number: MFCD00011198
CAS Number: 14285-68-8
UN: 3466
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR