Spiro[3.3]heptane-2-carboxylic acid ≥96%

Supplier: Ambeed
Warning

A363055-5G
77613-066EA 651.99 USD
77613-066
Spiro[3.3]heptane-2-carboxylic acid ≥96%
Spiro[3.3]heptane-2-carboxylic acid

Caution: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD19228416
CAS Number: 28114-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR