Nigericin sodium salt ≥98%
Supplier: Ambeed
|
Danger
|
A359670-100MG
A359670-10MG
A359670-1MG
A359670-50MG
77668-672EA
323.24
USD
77668-672
77668-674
77668-676
77668-678
Nigericin sodium salt ≥98%
Nigericin sodium salt
Caution: for Research Use Only
Formula:
C₄₀H₆₇NaO₁₁ |
MDL Number:
MFCD00036825 CAS Number: 28643-80-3 UN: 3462 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...