Saccharin sodium dihydrate ≥98%

Supplier: Ambeed
Warning

A361077-25G A361077-1000G A361077-100G A361077-500G
77668-680EA 7 USD
77668-680 77872-549 77872-550 77872-551
Saccharin sodium dihydrate ≥98%
Saccharin sodium dihydrate

Caution: for Research Use Only

Formula: C₇H₅NNaO₃S·2H₂O
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR