(2-Iodo-5-methoxyphenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A353101-250MG A353101-5G A353101-1G
77613-014EA 10.3 USD
77613-014 77686-005 77901-034
(2-Iodo-5-methoxyphenyl)boronic acid ≥95%
(2-Iodo-5-methoxyphenyl)boronic acid

Caution: for Research Use Only

Formula: C₇H₈BIO₃
MDL Number: MFCD26793796
CAS Number: 89694-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR