Nystatin 4840 IU/mg

Supplier: Ambeed
Warning

Synonyms: Fungicidin, Mycostatin

A326754-1G A326754-25G A326754-5G A326754-100G
77601-472EA 10.25 USD
77601-472 77601-474 77601-476 77668-466
Nystatin 4840 IU/mg
Nystatin

Caution: for Research Use Only

Formula: C₄₇H₇₅NO₁₇
MW: 926.09 g/mol
MDL Number: MFCD25541651
CAS Number: 1400-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR