Adenosine 5'-monophosphate sodium salt 98+%

Supplier: Ambeed
Warning

A310789-100G A310789-25G A310789-500G
77881-866EA 40 USD
77881-866 77881-867 77881-868
Adenosine 5'-monophosphate sodium salt 98+%
Adenosine 5'-monophosphate sodium salt

Caution: for Research Use Only

Formula: C₁₀H₁₄N₅NaO₇P
MDL Number: MFCD08689634
CAS Number: 13474-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR