Digitonin 50%

Supplier: Ambeed
Danger

Synonyms: Digitonin

A300453-100MG A300453-1G A300453-250MG A300453-50MG A300453-5G
77601-350EA 54.33 USD
77601-350 77601-352 77601-354 77601-356 77879-590
Digitonin 50%
Digitonin

Caution: for Research Use Only

Formula: C₅₆H₉₂O₂₉
MW: 1229.312 g/mol
MDL Number: MFCD00077729
CAS Number: 11024-24-1
UN: 3462
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR