Novobiocin sodium salt ≥95%

Supplier: Ambeed
Warning

Synonyms: Sodium 4-((7-(((2R,3R,4S,5R)-4-(carbamoyloxy)-3-hydroxy-5-methoxy-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-8-methyl-2-oxo-2H-chromen-3-yl)carbamoyl)-2-(3-methylbut-2-en-1-yl)phenolate, Novobiocin sodium

A299620-1MG A299620-5MG A299620-250MG A299620-1G A299620-100MG
77593-816EA 7.18 USD
77593-816 77601-342 77668-334 77668-332 77879-589
Novobiocin sodium salt ≥95%
Novobiocin sodium salt

Caution: for Research Use Only

Formula: C₃₁H₃₅N₂NaO₁₁
MDL Number: MFCD00066541
CAS Number: 1476-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR