1-Benzylpiperidine-4-carbaldehyde ≥95%

Supplier: Ambeed
Warning

A281868-1G A281868-100G A281868-25G A281868-5G
77571-750EA 16 USD
77571-750 77879-534 77879-535 77879-536
1-Benzylpiperidine-4-carbaldehyde ≥95%
1-Benzylpiperidine-4-carbaldehyde

Caution: for Research Use Only

Formula: C₁₃H₁₇NO
MDL Number: MFCD02258902
CAS Number: 22065-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR