Thiostrepton ≥98%

Supplier: Ambeed
Warning

A265605-10MG A265605-1MG A265605-250MG A265605-100MG A265605-50MG A265605-1G
77593-642EA 10.25 USD
77593-642 77593-644 77601-138 77668-142 77668-144 77872-300
Thiostrepton ≥98%
Thiostrepton

Caution: for Research Use Only

Formula: C₇₂H₈₅N₁₉O₁₈S₅
MDL Number: MFCD00135828
CAS Number: 1393-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR