SC-79 ≥98%

Supplier: Ambeed
Warning

A261312-250MG A261312-100MG A261312-50MG
77616-098EA 54.59 USD
77616-098 77685-743 77685-744
SC-79 ≥98%
SC-79

Caution: for Research Use Only

Formula: C₁₇H₁₇ClN₂O₅
MDL Number: MFCD02681303
CAS Number: 305834-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR