(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

A257946-100MG A257946-250MG
77601-078EA 27.68 USD
77601-078 77601-080
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥97%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

Caution: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
MDL Number: MFCD00064200
CAS Number: 17257-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR