4-((3S,5R,8R,9S,10S,12R,13S,14S,17R)-3,12,14-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)furan-2(5H)-one ≥98%

Supplier: Ambeed
Danger

A240397-1MG A240397-10MG
77626-274EA 25.75 USD
77626-274 77879-420
4-((3S,5R,8R,9S,10S,12R,13S,14S,17R)-3,12,14-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)furan-2(5H)-one ≥98%
4-((3S,5R,8R,9S,10S,12R,13S,14S,17R)-3,12,14-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)furan-2(5H)-one

Caution: for Research Use Only

Formula: C₂₃H₃₄O₅
CAS Number: 1672-46-4
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR