Cefpodoxime proxetil ≥95%

Supplier: Ambeed
Warning

Synonyms: (6R,7R)-1-((Isopropoxycarbonyl)oxy)ethyl 7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A234300-1G A234300-5MG A234300-5G
77600-882EA 36.9 USD
77600-882 77600-884 77626-242
Cefpodoxime proxetil ≥95%
Cefpodoxime proxetil

Caution: for Research Use Only

Formula: C₂₁H₂₇N₅O₉S₂
MW: 557.593 g/mol
MDL Number: MFCD00865088
CAS Number: 87239-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR