Methylenediphosphonic acid ≥98%

Supplier: Ambeed
Danger

A224115-1G A224115-250MG A224115-5G
77600-800EA 24.6 USD
77600-800 77667-862 77667-864
Methylenediphosphonic acid ≥98%
Methylenediphosphonic acid

Caution: for Research Use Only

Formula: CH₆O₆P₂
MDL Number: MFCD00002139
CAS Number: 1984-15-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR