MES sodium salt 98

Supplier: Ambeed
Warning

Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 2-morpholinoethane-1-sulfonate, Sodium 3-morpholin-1-ylethylsulphonate, Sodium 3-morpholin-1-ylethylsulfonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, Sodium 2-Morpholinoethanesulfonate, 2-(N-Morpholino)ethanesulfonic acid sodium salt, MES-Na

A1727886-100G A1727886-500G A1727886-25G
77625-808EA 27.81 USD
77625-808 77625-810 77877-202
MES sodium salt 98
MES sodium salt

Caution: for Research Use Only

Formula: C₆H₁₂NNaO₄S
MW: 217.22 g/mol
MDL Number: MFCD00065473
CAS Number: 71119-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR