β-Escin 97-103% (T), beta≥60%(HPLC), Ambeed.Inc (Loss on drying≤5%)

Supplier: Ambeed
Warning

A163721-1G A163721-5G A163721-250MG
77683-350EA 41 USD
77683-350 77683-351 77885-298
β-Escin 97-103% (T), beta≥60%(HPLC), Ambeed.Inc (Loss on drying≤5%)
β-Escin

Caution: For research use only.

MDL Number: MFCD00076054
CAS Number: 11072-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR