BI-D1870 ≥98%

Supplier: Ambeed
Warning

A153763-50MG A153763-100MG
77600-188EA 197.83 USD
77600-188 77871-762
BI-D1870 ≥98%
BI-D1870

Caution: for Research Use Only

Formula: C₁₉H₂₃F₂N₅O₂
MDL Number: MFCD11223662
CAS Number: 501437-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR