Eosin B ≥85% (dye content) BS

Supplier: Ambeed
Danger

Synonyms: 2,7-Dinitro-4,5-dibromofluoresceinEosin bluishSodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoateEosin B disodium saltAcid Red 912-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt

A1493870-25G A1493870-5G
77624-530EA 8.24 USD
77624-530 77597-686
Eosin B ≥85% (dye content) BS
Eosin B

Caution: for Research Use Only

Formula: C₂₀H₆Br₂N₂Na₂O₉
MW: 624.06 g/mol
MDL Number: MFCD00005041
CAS Number: 548-24-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR