(R)-N-((S)-1-(2-(Di(adamantan-1-yl)phosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide ≥95%

Supplier: Ambeed

A1487090-100MG A1487090-250MG A1487090-50MG
77597-604EA 235.75 USD
77597-604 77875-379 77875-380
(R)-N-((S)-1-(2-(Di(adamantan-1-yl)phosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide ≥95%
(R)-N-((S)-1-(2-(Di(adamantan-1-yl)phosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide

Caution: for Research Use Only

Formula: C₃₂H₄₈NOPS
MDL Number: MFCD33401811
CAS Number: 1929530-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR