mPEG-CHO, M.W. ~10000 ≥97%

Supplier: Ambeed
Warning

A1483102-100MG A1483102-1G A1483102-250MG
77875-305EA 53 USD
77875-305 77875-306 77875-307
mPEG-CHO, M.W. ~10000 ≥97%
mPEG-CHO

Caution: for Research Use Only

MDL Number: MFCD00803683
CAS Number: 125061-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR