mPEG-CHO, M.W. ~1000

Supplier: Ambeed
Warning

A1483051-100MG A1483051-250MG A1483051-1G
77597-560EA 39.98 USD
77597-560 77597-562 77624-366
mPEG-CHO, M.W. ~1000
mPEG-CHO

Caution: for Research Use Only

MDL Number: MFCD00803683
CAS Number: 125061-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR