(4S,7S,10S,13S)-4-Benzyl-13-(2-chloroacetyl)-10-((R)-1-hydroxyethyl)-7-isobutyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid ≥98%

Supplier: Ambeed
Danger

A1445371-1MG A1445371-5MG
77624-200EA 157.59 USD
77624-200 77874-890
(4S,7S,10S,13S)-4-Benzyl-13-(2-chloroacetyl)-10-((R)-1-hydroxyethyl)-7-isobutyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid ≥98%
(4S,7S,10S,13S)-4-Benzyl-13-(2-chloroacetyl)-10-((R)-1-hydroxyethyl)-7-isobutyl-2,5,8,11-tetraoxo-3,6,9,12-tetraazapentadecan-15-oic acid

Caution: for Research Use Only

Formula: C₂₆H₃₇ClN₄O₈
CAS Number: 2376255-48-8
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR