Polyethylene glycol 2,000

Supplier: Ambeed
Warning

Synonyms: PEG 2000PEG 2,000Ethylene glycol polymer 2000Ethylene glycol polymer average Mn 2000

A139063-100G A139063-1000G A139063-25G A139063-500G
77597-118EA 6.15 USD
77597-118 77597-116 77597-114 77597-120
Polyethylene glycol 2,000
Polyethylene glycol 2,000

Caution: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 2000 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR