2,2',2''-(10-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic acid ≥98%
Supplier: Ambeed
|
Warning
|
A1365476-25MG
A1365476-50MG
A1365476-100MG
A1365476-250MG
77867-600EA
141
USD
77867-600
77867-601
77867-598
77867-599
2,2',2''-(10-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic acid ≥98%
2,2',2''-(10-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic acid
Caution: for Research Use Only
Formula:
C₂₂H₃₄N₆O₉ |
MDL Number:
MFCD32708535 CAS Number: 1006711-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...