(((S)-5-Amino-1-carboxypentyl)carbamoyl)-L-glutamic acid compound with 2,2,2-trifluoroacetic acid (1:1) ≥98%

Supplier: Ambeed

A1355691-1MG A1355691-5MG
77596-866EA 11.28 USD
77596-866 77867-490
(((S)-5-Amino-1-carboxypentyl)carbamoyl)-L-glutamic acid compound with 2,2,2-trifluoroacetic acid (1:1) ≥98%
(((S)-5-Amino-1-carboxypentyl)carbamoyl)-L-glutamic acid compound with 2,2,2-trifluoroacetic acid (1:1)

Caution: for Research Use Only

Formula: C₁₄H₂₂F₃N₃O₉
MDL Number: MFCD32901379
CAS Number: 1269794-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR