Methyl-10-undecenoate ≥96%

Supplier: Ambeed
Danger

A129959-25G A129959-500G A129959-100G A129959-1000G
77599-882EA 12.3 USD
77599-882 77599-884 77666-860 77878-938
Methyl-10-undecenoate ≥96%
Methyl 10-undecenoate

Caution: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00016689
CAS Number: 111-81-9
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR