4-(Carboxymethyl)-3-fluorobenzeneboronic acid ≥98%

Supplier: Ambeed
Warning

A1241074-100MG A1241074-1G A1241074-250MG
77571-452EA 55 USD
77571-452 77866-948 77866-949
4-(Carboxymethyl)-3-fluorobenzeneboronic acid ≥98%
4-(Carboxymethyl)-3-fluorobenzeneboronic acid

Caution: for Research Use Only

Formula: C₈H₈BFO₄
MDL Number: MFCD32220289
CAS Number: 1146614-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR