4-Aminocinnamic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Aminophenyl)acrylic acid, 4-Aminocinnamic Acid, 3-(4-Aminophenyl)acrylic Acid

A122649-5G A122649-1G
77624-018EA 136.99 USD
77624-018 77876-888
4-Aminocinnamic acid ≥97%
4-Aminocinnamic acid

Caution: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00017117
CAS Number: 2393-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR