N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Antalarmin hydrochloride, N-Butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride, N-butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride, Antalarmin HCl

A1209598-10MG A1209598-1MG A1209598-5MG
77596-280EA 184.5 USD
77596-280 77596-282 77596-284
N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride ≥97%
N-Butyl-N-ethyl-7-mesityl-2,5,6-trimethyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride

Caution: for Research Use Only

Formula: C₂₄H₃₅ClN₄
MW: 415.01 g/mol
MDL Number: MFCD05664721
CAS Number: 220953-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR