(1R,2R)-2-(4-Bromophenyl)cyclopropane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A1181983-100MG A1181983-250MG
77866-405EA 224 USD
77866-405 77866-406
(1R,2R)-2-(4-Bromophenyl)cyclopropane-1-carboxylic acid ≥98%
(1R,2R)-2-(4-Bromophenyl)cyclopropane-1-carboxylic acid

Caution: for Research Use Only

Formula: C₁₀H₉BrO₂
MDL Number: MFCD17215399
CAS Number: 31501-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR