2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine ≥98%

Supplier: Ambeed
Warning

A1180254-250MG A1180254-100MG A1180254-1G
77596-176EA 116.85 USD
77596-176 77620-622 77620-624
2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine ≥98%
2,8-Diethyl-5,5-difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine

Caution: for Research Use Only

Formula: C₂₃H₂₇BF₂N₂
CAS Number: 189264-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR